Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otop2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172801.2 | NP_766389.2 | 4 - 5 | 776 | 77 | |
GenBank mRNA | AB087504.1 | - | 3 - 4 | 661 | 77 | |
AK028866.1 | - | 4 - 5 | 777 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |