Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpsap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026364.1 | NP_080640.1 | 6 - 7 | 765 | 91 | |
GenBank mRNA | AK011290.1 | - | 6 - 7 | 735 | 91 | |
AK077439.1 | - | 6 - 7 | 768 | 91 | ||
BC029621.1 | - | 6 - 7 | 756 | 4208743 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |