Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033528.1 | NP_001028700.1 | 4 - 5 | 874 | 80 | |
GenBank mRNA | AK028572.1 | - | 4 - 5 | 919 | 80 | |
AK049205.1 | - | 4 - 5 | 704 | 80 | ||
AK129363.1 | - | 4 - 5 | 672 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Protease Mus musculus Probe spans exons |