Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttyh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053273.2 | NP_444503.2 | 2 - 3 | 381 | 107 | |
GenBank mRNA | AK152172.1 | - | 2 - 3 | 333 | 107 | |
AK154835.1 | - | 2 - 3 | 389 | 107 | ||
AK155166.1 | - | 2 - 3 | 382 | 107 | ||
AK156393.1 | - | 2 - 3 | 334 | 107 | ||
AK168408.1 | - | 2 - 3 | 334 | 107 | ||
BC060740.1 | - | 2 - 3 | 324 | 4158813 | 107 | |
BY740853.1 | - | 2 - 3 | 361 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |