Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037809.5 | NP_001032898.1 | 13 - 14 | 2089 | 88 | |
NM_007665.3 | NP_031691.1 | 13 - 14 | 2086 | 88 | ||
GenBank mRNA | AK031265.1 | - | 13 - 14 | 2029 | 88 | |
AK045041.1 | - | 13 - 14 | 2049 | 88 | ||
AK164577.1 | - | 13 - 14 | 2059 | 88 | ||
BC052189.1 | - | 11 - 12 | 1777 | 88 | ||
BC098459.1 | - | 13 - 14 | 1995 | 30079668 | 88 | |
X06340.1 | - | 13 - 14 | 2027 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |