Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sin3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009188.4 | NP_033214.2 | 18 - 19 | 3106 | 60 | |
GenBank mRNA | AK013202.1 | - | 1 - 2 | 123 | 60 | |
AK087970.1 | - | 10 - 11 | 2483 | 60 | ||
AK159076.1 | - | 18 - 19 | 3089 | 60 | ||
AK173005.1 | - | 17 - 18 | 5056 | 60 | ||
CX211072.1 | - | 1 - 2 | 196 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Mus musculus Inventoried Probe spans exons |