Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276383.1 | NP_001263312.1 | 2 - 3 | 210 | 84 | |
NM_153060.3 | NP_694700.2 | 2 - 3 | 558 | 84 | ||
GenBank mRNA | AB441166.1 | - | 2 - 3 | 430 | 84 | |
AK156007.1 | - | 3 - 4 | 204 | 84 | ||
BC011467.1 | - | 2 - 3 | 558 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |