Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276383.1 | NP_001263312.1 | 7 - 8 | 863 | 77 | |
NM_153060.3 | NP_694700.2 | 7 - 8 | 1211 | 77 | ||
GenBank mRNA | AK156007.1 | - | 8 - 9 | 857 | 77 | |
BC011467.1 | - | 7 - 8 | 1211 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |