Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bbs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026116.2 | NP_080392.1 | 10 - 11 | 1684 | 99 | |
GenBank mRNA | AF342737.1 | - | 10 - 11 | 1684 | 99 | |
AK010779.1 | - | 10 - 11 | 1297 | 99 | ||
BC057184.1 | - | 10 - 11 | 1343 | 5066846 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |