Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153065.3 | NP_694705.2 | 17 - 18 | 2007 | 116 | |
GenBank mRNA | AK134274.1 | - | 17 - 18 | 2008 | 116 | |
AK138442.1 | - | 17 - 18 | 2031 | 116 | ||
AK146469.1 | - | 17 - 18 | 2035 | 116 | ||
BC011321.1 | - | 16 - 17 | 1884 | 4007058 | 116 | |
BC026381.1 | - | 2 - 3 | 196 | 116 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |