Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Susd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101510.1 | NP_001094980.1 | 1 - 2 | 285 | 139 | |
GenBank mRNA | AK132299.1 | - | 1 - 2 | 206 | 139 | |
BY718562.1 | - | 1 - 2 | 499 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |