Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nuak1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004363.1 | NP_001004363.1 | 2 - 3 | 475 | 87 | |
GenBank mRNA | AB182364.1 | - | 1 - 2 | 112 | 87 | |
BC082328.1 | - | 2 - 3 | 475 | 30355405 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |