Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197999.2 | NP_932116.2 | 2 - 3 | 356 | 70 | |
GenBank mRNA | AK146261.1 | - | 2 - 3 | 300 | 70 | |
BC027185.1 | - | 2 - 3 | 334 | 4194872 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |