Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0800009376.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009376.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009374.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009374.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009371.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009371.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009383.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009383.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009379.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009379.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009380.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009380.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009372.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009372.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009378.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009378.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009377.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009377.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009382.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009382.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009373.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009373.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009381.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009381.mm.1 |
TC0800001199.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800009375.mm.1 |
NM_001272045.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800009375.mm.1 |
TC0800001199.mm.1 |
MTA TC |