Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd11b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008289.2 | NP_032315.2 | 2 - 3 | 603 | 75 | |
GenBank mRNA | AK155153.1 | - | 2 - 3 | 605 | 75 | |
BC014753.1 | - | 2 - 3 | 594 | 4235545 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |