Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026030.2 | NP_080306.1 | 3 - 4 | 496 | 158 | |
GenBank mRNA | AK012338.1 | - | 3 - 4 | 409 | 158 | |
AK012817.1 | - | 2 - 3 | 286 | 158 | ||
AK076056.1 | - | 3 - 4 | 416 | 158 | ||
AK145401.1 | - | 3 - 4 | 401 | 158 | ||
AK145434.1 | - | 3 - 4 | 443 | 158 | ||
AK166862.1 | - | 3 - 4 | 400 | 158 | ||
BC003848.1 | - | 3 - 4 | 372 | 3594058 | 158 | |
BC100539.1 | - | 3 - 4 | 407 | 158 | ||
BC113766.1 | - | 3 - 4 | 368 | 40058503 | 158 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |