Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21769694 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21769693 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
588884 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
415782 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596117 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
164203 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
575925 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
217440 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
295310 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
605481 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
9016 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
571767 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839265 |
NM_009347.1 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
839265 |
X99805.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
88368 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
219796 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
64865 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
164184 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P134222 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P137506 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P252121 |
AK003786.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_52_P252121 |
AK003786.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769677 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769688 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769671 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769667 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769693 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769668 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769674 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769685 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769675 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769694 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769680 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P21769691 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00534779 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P134222 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00534772 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P137506 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04751343 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00534790 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315646 |
BC010586.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P315646 |
AJ222660.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P252121 |
AK003786.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P252121 |
AK003786.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P115330 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P137506 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P134222 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01887017 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01887006 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01887008 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01887022 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P01887019 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21769694 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P21769693 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769593 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769639 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769636 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769654 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769595 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769615 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769623 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769633 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769667 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769641 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769661 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769649 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769688 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769665 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769685 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769643 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769666 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769668 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769628 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769599 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769637 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769605 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769601 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769619 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769672 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769691 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769694 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769631 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769647 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769613 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769680 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769634 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769656 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769653 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21769693 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01887003 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769665 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01887006 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769685 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01177058 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02169795 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769668 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769680 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01177056 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769621 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P02159071 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01177055 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01151078 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769679 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01773517 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01828879 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769683 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769688 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769678 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769623 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01351725 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01197522 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769675 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P21769693 |
- |
- |
Agilent Six_ChIP_01_of_03_v2 |
A_68_P01887023 |
- |
- |