Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181734.3 | NP_859423.2 | 1 - 2 | 100 | 87 | |
GenBank mRNA | AK033600.1 | - | 1 - 2 | 49 | 87 | |
AK039450.1 | - | 1 - 2 | 101 | 87 | ||
AK052794.1 | - | 1 - 2 | 66 | 87 | ||
AK081680.1 | - | 1 - 2 | 63 | 87 | ||
AK156641.1 | - | 1 - 2 | 66 | 87 | ||
BC052389.1 | - | 1 - 2 | 59 | 5718874 | 87 | |
CJ183358.1 | - | 1 - 2 | 102 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |