Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vat1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173016.3 | NP_766604.2 | 5 - 6 | 891 | 105 | |
GenBank mRNA | AK122527.1 | - | 5 - 6 | 891 | 105 | |
AK189227.1 | - | 2 - 3 | 117 | 105 | ||
BC056927.1 | - | 5 - 6 | 901 | 5719211 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |