Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100038970.mm.1 |
NM_001033267.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038970.mm.1 |
NM_001033267.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038970.mm.1 |
TC1100004117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100038971.mm.1 |
NM_001033267.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038971.mm.1 |
NM_001033267.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038971.mm.1 |
TC1100004117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100038968.mm.1 |
NM_001033267.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038968.mm.1 |
NM_001033267.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038968.mm.1 |
TC1100004117.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100038969.mm.1 |
NM_001033267.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038969.mm.1 |
NM_001033267.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100038969.mm.1 |
TC1100004117.mm.1 |
MTA TC |