Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ten1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027107.1 | NP_081383.1 | 1 - 2 | 130 | 112 | |
GenBank mRNA | AK009156.1 | - | 1 - 2 | 97 | 112 | |
AK146257.1 | - | 1 - 2 | 120 | 112 | ||
AK153854.1 | - | 1 - 2 | 88 | 112 | ||
BC027639.1 | - | 1 - 2 | 75 | 1069092 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |