Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181549.3 | NP_853527.2 | 1 - 2 | 209 | 71 | |
GenBank mRNA | AK132888.1 | - | 3 - 4 | 1016 | 71 | |
AY223871.1 | - | 298 | 71 | |||
AY223873.1 | - | 269 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Detect Genomic DNA Mus musculus 1700 Probe spans exons |