Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178802.4 | NP_848917.2 | 1 - 2 | 539 | 141 | |
GenBank mRNA | AK028857.1 | - | 1 - 2 | 428 | 141 | |
AK047439.1 | - | 1 - 2 | 539 | 141 | ||
AK050080.1 | - | 1 - 2 | 451 | 141 | ||
AK079982.1 | - | 1 - 2 | 539 | 141 | ||
BC057094.1 | - | 428 | 6418691 | 141 | ||
BC118954.1 | - | 1 - 2 | 294 | 141 | ||
BC118955.1 | - | 1 - 2 | 294 | 141 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |