Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197959.2 | NP_932063.2 | 10 - 11 | 1562 | 74 | |
GenBank mRNA | AK156592.1 | - | 10 - 11 | 1532 | 74 | |
BC023289.1 | - | 4 - 5 | 523 | 74 | ||
BC049272.1 | - | 1 - 2 | 96 | 74 | ||
BC057614.1 | - | 10 - 11 | 1530 | 6401536 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |