Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif18b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_197959.2 | NP_932063.2 | 7 - 8 | 1217 | 56 | |
GenBank mRNA | AK156592.1 | - | 7 - 8 | 1187 | 56 | |
BC023289.1 | - | 1 - 2 | 178 | 56 | ||
BC057614.1 | - | 7 - 8 | 1185 | 6401536 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |