Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nploc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195023.1 | NP_001181952.1 | 14 - 15 | 1637 | 66 | |
GenBank mRNA | AK129375.1 | - | 14 - 15 | 1612 | 66 | |
AK141998.1 | - | 14 - 15 | 1636 | 66 | ||
AK149823.1 | - | 14 - 15 | 1609 | 66 | ||
AK149912.1 | - | 14 - 15 | 1636 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mus musculus Probe spans exons |