Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033194.2 | NP_001028366.1 | 15 - 16 | 2301 | 81 | |
GenBank mRNA | AK145040.1 | - | 2305 | 81 | ||
AK160997.1 | - | 15 - 16 | 2289 | 81 | ||
AK165830.1 | - | 15 - 16 | 2302 | 81 | ||
BC141157.1 | - | 15 - 16 | 2291 | 9055769 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |