Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028034.2 | NP_082310.1 | 3 - 4 | 529 | 89 | |
GenBank mRNA | AB211061.1 | - | 3 - 4 | 468 | 89 | |
AK010386.1 | - | 3 - 4 | 468 | 89 | ||
AK010714.1 | - | 2 - 3 | 320 | 89 | ||
AK135918.1 | - | 3 - 4 | 450 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |