Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fastkd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172422.3 | NP_766010.1 | 8 - 9 | 1702 | 146 | |
GenBank mRNA | AK035322.1 | - | 8 - 9 | 1702 | 146 | |
AK084971.1 | - | 8 - 9 | 1677 | 146 | ||
AK129251.1 | - | 6 - 7 | 2879 | 146 | ||
AK148224.1 | - | 1686 | 146 | |||
AK153916.1 | - | 7 - 8 | 1645 | 146 | ||
AK166378.1 | - | 8 - 9 | 1687 | 146 | ||
AK167714.1 | - | 7 - 8 | 1958 | 146 | ||
BC008271.1 | - | 7 - 8 | 1643 | 3603187 | 146 | |
BC057208.1 | - | 7 - 8 | 1636 | 5328766 | 146 | |
BC080813.1 | - | 7 - 8 | 1642 | 6394260 | 146 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |