Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Habp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146101.1 | NP_666213.1 | 5 - 6 | 497 | 74 | |
GenBank mRNA | AK128915.1 | - | 6 - 7 | 623 | 74 | |
AK132407.1 | - | 514 | 74 | |||
BC031775.1 | - | 5 - 6 | 497 | 4242577 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |