Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081264.1 | NP_001074733.1 | 10 - 11 | 1099 | 62 | |
GenBank mRNA | AK157903.1 | - | 10 - 11 | 1061 | 62 | |
AK171887.1 | - | 10 - 11 | 1052 | 62 | ||
BC050854.1 | - | 10 - 11 | 1064 | 6332345 | 62 | |
BC080867.1 | - | 10 - 11 | 1039 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |