Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmbim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027154.5 | NP_081430.3 | 3 - 4 | 554 | 71 | |
GenBank mRNA | AB097685.1 | - | 2 - 3 | 300 | 71 | |
AK077689.1 | - | 3 - 4 | 379 | 71 | ||
AK144110.1 | - | 3 - 4 | 398 | 71 | ||
AK147297.1 | - | 2 - 3 | 9262 | 71 | ||
AK152864.1 | - | 3 - 4 | 397 | 71 | ||
AK166926.1 | - | 3 - 4 | 570 | 71 | ||
AK170625.1 | - | 3 - 4 | 364 | 71 | ||
AK171102.1 | - | 3 - 4 | 396 | 71 | ||
AK220296.1 | - | 3 - 4 | 511 | 71 | ||
BC004752.1 | - | 2 - 3 | 344 | 3584518 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |