Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018868.2 | NP_061356.2 | 13 - 14 | 1571 | 173 | |
GenBank mRNA | AF053232.1 | - | 13 - 14 | 1476 | 173 | |
AK034577.1 | - | 225 | 174 | |||
AK044216.1 | - | 1568 | 173 | |||
AK134286.1 | - | 13 - 14 | 1572 | 173 | ||
BC076604.1 | - | 13 - 14 | 1572 | 30630088 | 173 | |
BC085135.1 | - | 13 - 14 | 1541 | 5718488 | 173 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |