Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018868.2 | NP_061356.2 | 2 - 3 | 289 | 145 | |
GenBank mRNA | AK044216.1 | - | 286 | 145 | ||
AK133959.1 | - | 2 - 3 | 287 | 145 | ||
AK134286.1 | - | 2 - 3 | 290 | 145 | ||
BC076604.1 | - | 2 - 3 | 290 | 30630088 | 145 | |
BC085135.1 | - | 2 - 3 | 259 | 5718488 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |