Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100003430.mm.1 |
NM_001008419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003430.mm.1 |
NM_001008419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003430.mm.1 |
TC0100000431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003432.mm.1 |
NM_001008419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003432.mm.1 |
NM_001008419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003432.mm.1 |
TC0100000431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003429.mm.1 |
NM_001008419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003429.mm.1 |
NM_001008419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003429.mm.1 |
TC0100000431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003433.mm.1 |
NM_001008419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003433.mm.1 |
NM_001008419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003433.mm.1 |
TC0100000431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003428.mm.1 |
NM_001008419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003428.mm.1 |
NM_001008419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003428.mm.1 |
TC0100000431.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100003431.mm.1 |
NM_001008419.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003431.mm.1 |
NM_001008419.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100003431.mm.1 |
TC0100000431.mm.1 |
MTA TC |