Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037737.2 | NP_001032826.1 | 15 - 16 | 1652 | 100 | |
NM_009709.4 | NP_033839.2 | 14 - 15 | 1607 | 100 | ||
GenBank mRNA | AK028546.1 | - | 14 - 15 | 1594 | 100 | |
AK040475.1 | - | 15 - 16 | 1654 | 100 | ||
AK141362.1 | - | 15 - 16 | 1523 | 100 | ||
AK145415.1 | - | 14 - 15 | 1562 | 100 | ||
AK152473.1 | - | 15 - 16 | 1634 | 100 | ||
AK153355.1 | - | 15 - 16 | 1634 | 100 | ||
AK220458.1 | - | 11 - 12 | 1783 | 100 | ||
BC012870.1 | - | 15 - 16 | 1614 | 3964932 | 100 | |
U10325.1 | - | 15 - 16 | 1516 | 100 | ||
U14333.1 | - | 14 - 15 | 1445 | 100 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |