Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201354.2 | NP_958742.2 | 7 - 8 | 920 | 94 | |
GenBank mRNA | BC058104.1 | - | 7 - 8 | 778 | 6833739 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |