Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdelc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023645.3 | NP_076134.1 | 3 - 4 | 869 | 87 | |
GenBank mRNA | AJ404004.1 | - | 3 - 4 | 631 | 87 | |
AK007835.1 | - | 3 - 4 | 862 | 87 | ||
AK035391.1 | - | 3 - 4 | 862 | 87 | ||
AK035899.1 | - | 3 - 4 | 870 | 87 | ||
AK053315.1 | - | 3 - 4 | 862 | 87 | ||
AK079701.1 | - | 3 - 4 | 862 | 87 | ||
AK170300.1 | - | 3 - 4 | 898 | 87 | ||
BC023141.1 | - | 3 - 4 | 849 | 4018698 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |