Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007408.3 | NP_031434.3 | 1 - 2 | 115 | 82 | |
GenBank mRNA | AK002911.1 | - | 1 - 2 | 51 | 82 | |
AK090277.1 | - | 1 - 2 | 46 | 82 | ||
AK150177.1 | - | 1 - 2 | 52 | 82 | ||
AK152081.1 | - | 1 - 2 | 51 | 82 | ||
AK152263.1 | - | 1 - 2 | 43 | 82 | ||
AK152877.1 | - | 1 - 2 | 51 | 82 | ||
AK152896.1 | - | 1 - 2 | 43 | 82 | ||
AK152982.1 | - | 1 - 2 | 74 | 82 | ||
AK153222.1 | - | 1 - 2 | 73 | 82 | ||
AK154582.1 | - | 1 - 2 | 52 | 82 | ||
AK167140.1 | - | 1 - 2 | 53 | 82 | ||
BC047124.1 | - | 1 - 2 | 39 | 82 | ||
BY297427.1 | - | 1 - 2 | 116 | 82 | ||
M93275.1 | - | 1 - 2 | 52 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |