Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007408.3 | NP_031434.3 | 2 - 3 | 168 | 54 | |
GenBank mRNA | AK002911.1 | - | 2 - 3 | 104 | 54 | |
AK090277.1 | - | 2 - 3 | 99 | 54 | ||
AK150177.1 | - | 2 - 3 | 105 | 54 | ||
AK152081.1 | - | 2 - 3 | 104 | 54 | ||
AK152263.1 | - | 2 - 3 | 96 | 54 | ||
AK152877.1 | - | 2 - 3 | 104 | 54 | ||
AK152896.1 | - | 2 - 3 | 96 | 54 | ||
AK152982.1 | - | 2 - 3 | 127 | 54 | ||
AK153222.1 | - | 2 - 3 | 126 | 54 | ||
AK154582.1 | - | 2 - 3 | 105 | 54 | ||
AK167140.1 | - | 2 - 3 | 106 | 54 | ||
AY035850.1 | - | 1 - 2 | 27 | 54 | ||
AY035851.1 | - | 1 - 2 | 27 | 54 | ||
BC047124.1 | - | 2 - 3 | 92 | 54 | ||
BC054766.1 | - | 2 - 3 | 81 | 6414796 | 54 | |
BY297427.1 | - | 2 - 3 | 169 | 54 | ||
M93275.1 | - | 2 - 3 | 105 | 54 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |