Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017427.1 | NP_001017427.1 | 7 - 8 | 835 | 114 | |
GenBank mRNA | AK132691.1 | - | 7 - 8 | 815 | 114 | |
BC030882.1 | - | 3 - 4 | 232 | 114 | ||
BC040766.1 | - | 3 - 4 | 221 | 114 | ||
BC125539.1 | - | 6 - 7 | 649 | 40130054 | 114 | |
BC137774.1 | - | 6 - 7 | 649 | 8860793 | 114 | |
BC144901.1 | - | 6 - 7 | 649 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |