Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022996.1 | NP_075372.1 | 1 - 2 | 207 | 151 | |
GenBank mRNA | AF220209.1 | - | 1 - 2 | 245 | 151 | |
AK050560.1 | - | 1 - 2 | 214 | 151 | ||
BC020359.1 | - | 1 - 2 | 194 | 151 | ||
BC026372.1 | - | 1 - 2 | 207 | 4162070 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |