Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tars2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163617.1 | NP_001157089.1 | 8 - 9 | 1069 | 69 | |
NM_001163618.1 | NP_001157090.1 | 6 - 7 | 826 | 69 | ||
NM_001163619.1 | NP_001157091.1 | 8 - 9 | 1069 | 69 | ||
NM_027931.3 | NP_082207.2 | 8 - 9 | 1069 | 69 | ||
GenBank mRNA | AK011540.1 | - | 8 - 9 | 955 | 69 | |
AK080690.1 | - | 8 - 9 | 977 | 69 | ||
AK088675.1 | - | 7 - 8 | 819 | 69 | ||
AK142685.1 | - | 8 - 9 | 964 | 69 | ||
AK164589.1 | - | 8 - 9 | 976 | 69 | ||
AK164825.1 | - | 8 - 9 | 848 | 69 | ||
AK168075.1 | - | 6 - 7 | 720 | 69 | ||
AK199920.1 | - | 4 - 5 | 330 | 69 | ||
BC008995.1 | - | 8 - 9 | 954 | 3157108 | 69 | |
BC067392.1 | - | 2 - 3 | 2177 | 69 | ||
BC147501.1 | - | 8 - 9 | 953 | 9056383 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |