Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Malt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172833.2 | NP_766421.1 | 10 - 11 | 1567 | 80 | |
GenBank mRNA | AK041919.1 | - | 10 - 11 | 1572 | 80 | |
AK085071.1 | - | 10 - 11 | 1567 | 80 | ||
BC110487.1 | - | 11 - 12 | 1574 | 40048907 | 80 | |
BC110488.1 | - | 10 - 11 | 1541 | 40048908 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Protease Mammalian Gene Collection Mus musculus Probe spans exons |