Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ablim3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164491.1 | NP_001157963.1 | 8 - 9 | 1025 | 83 | |
NM_198649.3 | NP_941051.2 | 7 - 8 | 1344 | 83 | ||
GenBank mRNA | AK086538.1 | - | 7 - 8 | 1124 | 83 | |
AK173040.1 | - | 7 - 8 | 902 | 83 | ||
BC060275.1 | - | 8 - 9 | 996 | 6822626 | 83 | |
BC094229.1 | - | 7 - 8 | 1318 | 30534236 | 83 | |
CF171696.1 | - | 2 - 3 | 120 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |