Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008985.2 | NP_033011.2 | 2 - 3 | 258 | 123 | |
GenBank mRNA | AK041296.1 | - | 2 - 3 | 264 | 123 | |
AK076223.1 | - | 2 - 3 | 237 | 123 | ||
AK139398.1 | - | 2 - 3 | 258 | 123 | ||
AK163141.1 | - | 2 - 3 | 234 | 123 | ||
AK220467.1 | - | 2 - 3 | 216 | 123 | ||
BB869850.1 | - | 2 - 3 | 202 | 123 | ||
BC068165.1 | - | 2 - 3 | 190 | 123 | ||
DQ832283.1 | - | 2 - 3 | 235 | 123 | ||
U11812.1 | - | 2 - 3 | 182 | 123 | ||
X74438.1 | - | 2 - 3 | 275 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |