Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfand2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159905.1 | NP_001153377.1 | 6 - 7 | 611 | 63 | |
NM_001159906.1 | NP_001153378.1 | 6 - 7 | 608 | 63 | ||
NM_026846.3 | NP_081122.2 | 5 - 6 | 645 | 63 | ||
GenBank mRNA | AK151430.1 | - | 5 - 6 | 647 | 63 | |
BC011495.1 | - | 5 - 6 | 670 | 4196340 | 63 | |
CX217579.1 | - | 6 - 7 | 614 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |