Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177222.4 | NP_796196.3 | 10 - 11 | 1264 | 71 | |
GenBank mRNA | AK132912.1 | - | 10 - 11 | 1488 | 71 | |
AY423542.1 | - | 10 - 11 | 1195 | 71 | ||
AY485607.1 | - | 10 - 11 | 1264 | 71 | ||
AY485608.1 | - | 10 - 11 | 1264 | 71 | ||
BC072568.1 | - | 8 - 9 | 1106 | 71 | ||
BC120743.1 | - | 10 - 11 | 1168 | 40129666 | 71 | |
BC125423.1 | - | 10 - 11 | 1168 | 40129938 | 71 | |
BC144801.1 | - | 10 - 11 | 1168 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |