Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026549.3 | NP_080825.1 | 1 - 2 | 113 | 65 | |
GenBank mRNA | AK003339.1 | - | 1 - 2 | 122 | 65 | |
AK077948.1 | - | 1 - 2 | 114 | 65 | ||
AK151495.1 | - | 1 - 2 | 117 | 65 | ||
AK154811.1 | - | 1 - 2 | 129 | 65 | ||
BC115797.1 | - | 1 - 2 | 112 | 40103840 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |