Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC096484.1 | - | 1 - 2 | 648 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |